Learn More
Medchemexpress LLC Lycopene | 502-65-8 | 98.4% | 536.87 | 50 MG
SDP
Catalog No. 5000427221
Encompass_Preferred
Lycopene is a naturally occurring carotenoid found in tomatoes, tomato products, and other red fruits and vegetables. It exhibits antioxidant effects.
  • Naturally occurring carotenoid
  • Exhibits antioxidant effects
  • More than 15 cited publications
Catalog No. 50-004-27221 Supplier Medchemexpress LLC Supplier No. HYN028750MG
Lycopene is a naturally occurring carotenoid found in tomatoes, tomato products, and other red fruits and vegetables. It exhibits antioxidant effects.
  • Naturally occurring carotenoid
  • Exhibits antioxidant effects
  • More than 15 cited publications