Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lycopene, >90%, MP Biomedicals™

Catalog No. ICN19868405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19868405 5 mg
ICN19868401 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19868405 Supplier MP Biomedicals, Inc Supplier No. 0219868405
Add to Cart
Add to Cart

Chemical Identifiers

CAS 502-65-8
Molecular Formula C40H56
Molecular Weight (g/mol) 536.888
InChI Key OAIJSZIZWZSQBC-GYZMGTAESA-N
Synonym lycopene, psi,psi-carotene, all-trans-lycopene, trans-lycopene, lycopene 7, lycored, lycopene van, lycopene, all-trans, unii-sb0n2n0wv6, ccris 7925
PubChem CID 446925
ChEBI CHEBI:15948
IUPAC Name (6E,8E,10E,12E,14E,16E,18E,20E,22E,24E,26E)-2,6,10,14,19,23,27,31-octamethyldotriaconta-2,6,8,10,12,14,16,18,20,22,24,26,30-tridecaene
SMILES CC(=CCCC(=CC=CC(=CC=CC(=CC=CC=C(C)C=CC=C(C)C=CC=C(C)CCC=C(C)C)C)C)C)C

Specifications

Color Red
Quantity 5 mg
Formula Weight 536.9
Percent Purity >90%
Physical Form Crystalline Needles
Chemical Name or Material Lycopene
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.