Learn More
Sigma Aldrich Fine Chemicals Biosciences Lycopene 98 HPLC from toma1MG
Catalog No. 502897317
Encompass
Lycopene is a carotenoid found in many red fruits and vegetables such as watermelon pink guava and tomato. Lycopene is a potent antioxidant. It is suggested that lycopene is used by the plant to protect against reactive oxygen species (ROS).
Catalog No. 50-289-7317 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SMB007061MG
Lycopene is a carotenoid found in many red fruits and vegetables such as watermelon pink guava and tomato. Lycopene is a potent antioxidant. It is suggested that lycopene is used by the plant to protect against reactive oxygen species (ROS).