Learn More
Invitrogen™ Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), DyLight 488
Description
The tomato lectin LEL is widely used as marker of blood vessels and microglial cells in rodents and in neuroscience research. It is a valuable tool in studies of rodent tumor angiogenesis and the tracing of neovascular development in xenograft models and brain research. LEL-DyLight 488 can be applied directly to tissue sections, or it can be introduced through intravascular perfusion for fluorescence detection.
- Very bright DyLight 488 conjugate detected with traditional FITC filter set, Ex/Em: 493/518 nm
- Applications: immunofluorescence, glycobiology
- Sugar specificity: [GlcNAc] 1,3-N-acetylglucosamine
- Inhibiting/eluting sugar: chitin hydrolysate
- Blood group specificity: non-specific
- Unconjugated lectin molecular weight: 71 KDa.
- Immunofluorescence working concentration: 5–20 μg/mL
LEL is a very stable single-subunit glycoprotein containing ∼50 percent arabinose and galactose and may form multimeric aggregates in solution. It is provided at a 1 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+. If a precipitate forms upon long-term storage, warm to 37°C.
Specifications
Specifications
| Quantity | 1 mg |
| Source | Tomato |
| Conjugate | DyLight 488 |
| Content And Storage | When stored at or below 2–8°C in the dark, the product is stable for 6 months after receipt. |
| Label Type | DyLight Fluors |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
| Form | Solution |
| Protein Subtype | Lectins |
| Product Line | DyLight |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.