Learn More
Invitrogen™ Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), Texas Red
Description
- Lycopersicon esculentum (Tomato) Lectin (LEL, TL), Texas Red, is a bright-red LEL-fluorophore conjugate. The tomato lectin LEL is widely used as marker of blood vessels and microglial cells in rodents and in neuroscience research.
- It is a valuable tool in studies of rodent tumor angiogenesis and the tracing of neovascular development in xenograft models and brain research.
- LEL-Texas Red can be applied directly to tissue sections, or it can be introduced through intravascular perfusion for fluorescence detection.
- Very bright Texas Red conjugate detected with traditional FITC filter set, Ex/Em: 595/615 nm
- Applications: immunofluorescence, glycobiology
- Sugar specificity: [GlcNAc] 1,3-N-acetylglucosamine
- Inhibiting/eluting sugar: chitin hydrolysate
- Blood group specificity: non-specific
- Unconjugated lectin molecular weight: 71 KDa.
- Immunofluorescence working concentration: 5–20 μg/mL
- LEL is a very stable single-subunit glycoprotein containing ∼50 percent arabinose and galactose and may form multimeric aggregates in solution.
- It is provided at a 1 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+. If a precipitate forms upon long-term storage, warm to 37°C.
Specifications
Specifications
| Quantity | 1 mg |
| Source | Tomato |
| Conjugate | Texas Red |
| Content And Storage | When stored at or below 2–8°C in the dark, the product is stable for 6 months after reciept. |
| Form | Solution |
| Protein Subtype | Lectins |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.