Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lycopodium Powder, Reagents

Catalog No. 01337193
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-193 25 g
01-337-194 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-193 Supplier Reagents Holdings Llc Supplier No. C223600025C1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lycopodium Powder, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 8023-70-9
Synonym Club moss spores

Specifications

Color Yellowish
Packaging Glass Bottle
Quantity 25 g
Specific Gravity 0.26
Grade Reagent
Physical Form Solid
Chemical Name or Material Lycopodium Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.