Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lycopodium, Powder, Spectrum™ Chemical
SDP

Catalog No. 18611397 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-397 50 kg Fiber Drum
18-611-393 12 kg Fiber Drum
18-611-395 2.5 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-611-397 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. LY10150KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 8023-70-9

Specifications

CAS Max % 1
Packaging Fiber Drum
Quantity 50 kg
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Lycopodium, Powder

CAUTION: For professional use only. Not for household use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.