Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lycoramine | 21133-52-8 | 99.8% | 289.37 g/mol | C17H23NO3 | 10 MG
SDP

Catalog No. 5000289690
Encompass_Preferred

Lycoramine is a dihydro-derivative of galanthamine isolated from Lycoris radiata. It functions as a potent acetylcholinesterase (AChE) inhibitor and is supplied for research use in biochemical and pharmacological studies. Refer to the manufacturer's documentation for COA and SDS.

  • Potent acetylcholinesterase inhibitor used in in vitro assays.
  • Available as a solid powder suitable for solution preparation.
  • High purity (~99.8%), with certificate of analysis available.
  • Applicable to neuroscience and cholinergic pathway research.
  • Documented chemical identifiers: CAS 21133-52-8; formula C17H23NO3.

Catalog No. 50-002-89690 Supplier Medchemexpress LLC Supplier No. HYN6619A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lycoramine is a dihydro-derivative of galanthamine isolated from Lycoris radiata. It functions as a potent acetylcholinesterase (AChE) inhibitor and is supplied for research use in biochemical and pharmacological studies. Refer to the manufacturer's documentation for COA and SDS.

  • Potent acetylcholinesterase inhibitor used in in vitro assays.
  • Available as a solid powder suitable for solution preparation.
  • High purity (~99.8%), with certificate of analysis available.
  • Applicable to neuroscience and cholinergic pathway research.
  • Documented chemical identifiers: CAS 21133-52-8; formula C17H23NO3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.