Learn More
Selleck Chemical LLC Lycorine-25mg
SDP
Catalog No. 502389664
Encompass_Preferred
Lycorine (Galanthidine, Amarylline, Narcissine, Licorine, Belamarine) is a toxic crystalline alkaloid found in various Amaryllidaceae species that weakly inhibits acetylcholinesterase (AChE) and ascorbic acid biosynthesis.
Catalog No. 50-238-9664 Supplier Selleck Chemical LLC Supplier No. S390325MG
Lycorine (Galanthidine, Amarylline, Narcissine, Licorine, Belamarine) is a toxic crystalline alkaloid found in various Amaryllidaceae species that weakly inhibits acetylcholinesterase (AChE) and ascorbic acid biosynthesis.