Learn More
Apexbio Technology LLC LYCORINE-CHLORIDE-20MG
SDP
Catalog No. 5000567777
Encompass_Preferred
Lycorine chloride (CAS No 2188-68-3) is a natural alkaloid derived from the Amaryllidaceae plant family frequently studied in the field of biomedical research for its diverse biological activities Its mechanism of action involves the inhibition of protein synthesis specifically targeting ribosomal pathways and interfering with viral replication processes making it of particular interest in antiviral research In vitro studies have demonstrated that lycorine chloride exhibits activity in the nanomolar to low micromolar range It is commonly utilized in cell models such as cancer cells and virus-infected cells as well as in animal studies to explore its potential therapeutic effects The compound is valuable in drug screening projects due to its ability to modulate biological pathways efficiently Experimental concentrations typically depend on the specific research design allowing for tailoring to particular study needs and objectives
Catalog No. 50-005-67777 Supplier Apexbio Technology LLC Supplier No. N155520
Lycorine chloride (CAS No 2188-68-3) is a natural alkaloid derived from the Amaryllidaceae plant family frequently studied in the field of biomedical research for its diverse biological activities Its mechanism of action involves the inhibition of protein synthesis specifically targeting ribosomal pathways and interfering with viral replication processes making it of particular interest in antiviral research In vitro studies have demonstrated that lycorine chloride exhibits activity in the nanomolar to low micromolar range It is commonly utilized in cell models such as cancer cells and virus-infected cells as well as in animal studies to explore its potential therapeutic effects The compound is valuable in drug screening projects due to its ability to modulate biological pathways efficiently Experimental concentrations typically depend on the specific research design allowing for tailoring to particular study needs and objectives