Learn More
Enzo Life Sciences Lycorine. HCl (10mg). CAS: 2188-68-3
SDP
Supplier:  Enzo Life Sciences BMLGR3130010
Encompass_Preferred
Lycorine is a eukaryotic termination inhibitor. Purity: ≥97% (Assay). Solubility: Soluble in water (>15mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0964