Learn More
Selleck Chemical LLC Lycorine hydrochloride-25mg
SDP
Catalog No. 502389984
Encompass_Preferred
Lycorine (Galanthidine, Amarylline, Narcissine, Licorine), a natural alkaloid extracted from the Amaryllidaceae plant family, has been reported to exhibit a wide range of physiological effects, including the potential effect against cancer. It is an HCV inhibitor with strong activity.
Catalog No. 50-238-9984 Supplier Selleck Chemical LLC Supplier No. S380025MG
Lycorine (Galanthidine, Amarylline, Narcissine, Licorine), a natural alkaloid extracted from the Amaryllidaceae plant family, has been reported to exhibit a wide range of physiological effects, including the potential effect against cancer. It is an HCV inhibitor with strong activity.