Learn More
Cayman Chemical LydIcamycIn 2.5mg
SDP
Catalog No. 502981521
Encompass_Preferred
An antibiotic that is particularly cytotoxic against S. aureus, including the methicillin resistant 535 strain (MICs = 3.1-6.2 UG/ml)
Catalog No. 50-298-1521 Supplier Cayman Chemical Supplier No. 205842.5 MG
An antibiotic that is particularly cytotoxic against S. aureus, including the methicillin resistant 535 strain (MICs = 3.1-6.2 UG/ml)