Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ LYFO DISK™ Staphylococcus epidermidis derived from ATCC™ 35984™†
SDP

Catalog No. 23017272 Shop All Microbiologics Products
Change view
Click to view available options
ATCC®:
ATCC™ 35984™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. ATCC®
23-017-272 ATCC™ 35984™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-017-272 Supplier Microbiologics™ Supplier No. 01189L
Add to Cart
Add to Cart
This item is not returnable. View return policy

For microbial identification

Specifications

ATCC® ATCC™ 35984™
Biosafety Level 1
Description LYFO DISK™ Staphylococcus epidermidis derived from ATCC™ 35984™
Detects Staphylococcus epidermidis derived from ATCC™ 35984™
Format LYFO DISK™
For Use With (Application) Microbial Identification
Incubation Time 24 to 48 hr.

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.