Learn More
Medchemexpress LLC Lyg-409 | 3053857-55-6 | 98.1% | 515.44 | 1 ML
SDP
Catalog No. 5000386499
Encompass_Preferred
LYG-409 is an orally active degrader of GSPT1. It demonstrates excellent anti-acute myeloid leukemia and prostate cancer activity in vivo, with TGI of 94.34% and 104.49%, respectively. It inhibits KG-1 cells by degrading GSPT1, with an IC50 of 9.50 nM and a DC50 of 7.87 nM in vitro.
  • Orally active degrader of GSPT1
  • Excellent anti-acute myeloid leukemia and prostate cancer activity in vivo
  • Inhibits KG-1 cells mediated by GSPT1 degradation
Catalog No. 50-003-86499 Supplier Medchemexpress LLC Supplier No. HY17095210MM/1ML
LYG-409 is an orally active degrader of GSPT1. It demonstrates excellent anti-acute myeloid leukemia and prostate cancer activity in vivo, with TGI of 94.34% and 104.49%, respectively. It inhibits KG-1 cells by degrading GSPT1, with an IC50 of 9.50 nM and a DC50 of 7.87 nM in vitro.
  • Orally active degrader of GSPT1
  • Excellent anti-acute myeloid leukemia and prostate cancer activity in vivo
  • Inhibits KG-1 cells mediated by GSPT1 degradation