Learn More
Medchemexpress LLC LYG-409 | 3053857-55-6 | 98.1% | 515.44 | 100 MG
SDP
Catalog No. 5000386179
Encompass_Preferred
LYG-409 is an orally active degrader of GSPT1. It has demonstrated excellent anti-acute myeloid leukemia and prostate cancer activity in vivo, with Tumor Growth Inhibition (TGI) rates of 94.34% and 104.49%, respectively. In vitro, it inhibits KG-1 cells through the degradation of GSPT1, exhibiting an IC50 of 9.50 nM and a DC50 of 7.87 nM.
  • Orally active GSPT1 degrader.
  • Shows excellent anti-acute myeloid leukemia and prostate cancer effects in vivo.
  • Inhibits KG-1 cells through GSPT1 degradation.
  • For research use only.
Catalog No. 50-003-86179 Supplier Medchemexpress LLC Supplier No. HY170952100MG
LYG-409 is an orally active degrader of GSPT1. It has demonstrated excellent anti-acute myeloid leukemia and prostate cancer activity in vivo, with Tumor Growth Inhibition (TGI) rates of 94.34% and 104.49%, respectively. In vitro, it inhibits KG-1 cells through the degradation of GSPT1, exhibiting an IC50 of 9.50 nM and a DC50 of 7.87 nM.
  • Orally active GSPT1 degrader.
  • Shows excellent anti-acute myeloid leukemia and prostate cancer effects in vivo.
  • Inhibits KG-1 cells through GSPT1 degradation.
  • For research use only.