Learn More
Medchemexpress LLC LYG-409 GSPT1 degrader | 3053857-55-6 | 98.1% | 515.44 | 50 MG
SDP
Catalog No. 5000386500
Encompass_Preferred
LYG-409 is an orally active degrader of GSPT1. It demonstrates excellent anti-acute myeloid leukemia and prostate cancer activity in vivo, with TGI of 94.34% and 104.49% respectively. LYG-409 inhibits KG-1 cells through the degradation of GSPT1 with an IC50 of 9.50 nM, and a DC50 of 7.87 nM in vitro.
  • Orally active degrader of GSPT1.
  • Exhibits anti-acute myeloid leukemia and prostate cancer activity in vivo.
  • Inhibits KG-1 cells by degrading GSPT1.
Catalog No. 50-003-86500 Supplier Medchemexpress LLC Supplier No. HY17095250MG
LYG-409 is an orally active degrader of GSPT1. It demonstrates excellent anti-acute myeloid leukemia and prostate cancer activity in vivo, with TGI of 94.34% and 104.49% respectively. LYG-409 inhibits KG-1 cells through the degradation of GSPT1 with an IC50 of 9.50 nM, and a DC50 of 7.87 nM in vitro.
  • Orally active degrader of GSPT1.
  • Exhibits anti-acute myeloid leukemia and prostate cancer activity in vivo.
  • Inhibits KG-1 cells by degrading GSPT1.