Learn More
Aviva Systems Biology Corp LYG2 Antibody - middle region (ARP60959_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60959_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LYG2. It was validated on Western Blot. . Alternate names: LYGH, MGC119046, MGC119047, MGC119049ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-213