Learn More
ATCC LYI Entamoeba Medium 1 vial prepares 15 ml of medium
Catalog No. 502382656
Encompass
This medium is the recommended formulation for the axenic cultivation of Entamoeba, Hypotrichomonas, Mastigamoeba, Pentatrichomonas, Tetratrichomonas, Trichomitus, Trichomonas, and Tritrichomonas. It is provided freeze-dried, complete with 10% (1.5 mL) bovine serum, 1 vial prepares 15 mL of medium.
Catalog No. 50-238-2656 Supplier ATCC Supplier No. PRA2154
This medium is the recommended formulation for the axenic cultivation of Entamoeba, Hypotrichomonas, Mastigamoeba, Pentatrichomonas, Tetratrichomonas, Trichomitus, Trichomonas, and Tritrichomonas. It is provided freeze-dried, complete with 10% (1.5 mL) bovine serum, 1 vial prepares 15 mL of medium.