Learn More
Aviva Systems Biology Corp LYL1 Antibody - C-terminal region (ARP31393_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31393_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LYL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LYL1. Alternate names: bHLHa18ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-54204