Learn More
Cederlane LYMPHOCYTE CELL SEPARATION MED
SDP
Catalog No. NC9284844
Encompass
Lympholyte�-M Cell Separation Media (Isolation of lymphocytes from Mouse lymphoid tissue), sterile liquid, Size: 5x30ml
Catalog No. NC9284844 Supplier Cederlane Supplier No. CL5030
Lympholyte�-M Cell Separation Media (Isolation of lymphocytes from Mouse lymphoid tissue), sterile liquid, Size: 5x30ml