Learn More
Promocell GmbH PromoCell Lymphocyte Separation Medium 1077 500 ml
Catalog No. 50306411
Encompass
The PromoCell Lymphocyte Separation Medium 1077 is intended for the separation of the portion of vital mononuclear cells from whole blood, buffy coats, bone marrow and several other starting materials, e.g. crude cell preparations, by means of low density gradient centrifugation. Lymphocyte separation Medium has a density of 1.077 g/ml at 20°C. It is sterile, ready-to use and based on Ficoll 400 and sodium diatrizoate providing optimal physiological parameters and low cytotoxicity.
Catalog No. 50-306-411 Supplier Promocell GmbH Supplier No. C44010
The PromoCell Lymphocyte Separation Medium 1077 is intended for the separation of the portion of vital mononuclear cells from whole blood, buffy coats, bone marrow and several other starting materials, e.g. crude cell preparations, by means of low density gradient centrifugation. Lymphocyte separation Medium has a density of 1.077 g/ml at 20°C. It is sterile, ready-to use and based on Ficoll 400 and sodium diatrizoate providing optimal physiological parameters and low cytotoxicity.