Learn More
Corning™ Lymphocyte Serum-free Medium, KBM 551
Catalog No. MT88551CM Shop All Corning Products
Change view
Click to view available options
Quantity:
1000 mL
Catalog No. Quantity
MT88551CM 1000 mL
1 options
This item is not returnable. View return policy
Catalog No. MT88551CM Supplier Corning™ Supplier No. 88551CM
Add to Cart
Add to Cart
This item is not returnable. View return policy
The use of T-lymphocytes in adaptive immunotherapy is emerging as an important tool in the treatment of cancer and other clinical applications.
Corning’s™ Lymphocyte Serum Free Medium (88-551-CM) supports the proliferation and activation of lymphocytes, including tumor infiltrating lymphocytes (TIL), Cytokine-Induced Killer (CIK) cells and Natural Killer (NK) cells. There is technical data to support the use in CAR-T as well as several other important immunotherapy applications.
TRUSTED_SUSTAINABILITY
Specifications
Form Liquid
Quantity 1000 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov