Learn More
Cederlane Lympholyte-H Cell Separation Media (Isolation of lymphocytes from Human peripheral blood), liquid (IVD) | 100mL
SDP
Catalog No. NC1790629
Encompass
Lympholyte-H can be utilized with a simple protocol for the elimination of erythrocytes and dead cells from human blood. Lympholyte-H also removes the majority of granulocytes (including neutrophils). The resulting cell population consists of a high and non-selective recovery of viable human lymphocytes and monocytes.
Catalog No. NC1790629 Supplier Cederlane Supplier No. CL5015
Lympholyte-H can be utilized with a simple protocol for the elimination of erythrocytes and dead cells from human blood. Lympholyte-H also removes the majority of granulocytes (including neutrophils). The resulting cell population consists of a high and non-selective recovery of viable human lymphocytes and monocytes.