Learn More
Cederlane LYMPHOLYTE-H 500ML
SDP
Catalog No. NC0631008
Encompass
Lympholyte�-H Cell Separation Media (Isolation of lymphocytes from Human peripheral blood), sterile liquid, Size: 500ml
Catalog No. NC0631008 Supplier Cederlane Supplier No. CL5020
Lympholyte�-H Cell Separation Media (Isolation of lymphocytes from Human peripheral blood), sterile liquid, Size: 500ml