Learn More
Cederlane LYMPHOLYTE-M 500ML
SDP
Catalog No. NC9990624
Encompass
Lympholyte�-M Cell Separation Media (Isolation of lymphocytes from Mouse lymphoid tissue), sterile liquid, Size: 500ml
Catalog No. NC9990624 Supplier Cederlane Supplier No. CL5035
Lympholyte�-M Cell Separation Media (Isolation of lymphocytes from Mouse lymphoid tissue), sterile liquid, Size: 500ml