Learn More
Cederlane LYMPHOLYTE-MAMMAL 100ML
SDP
Catalog No. NC9025455
Encompass
Lympholyte;-Mammal; Sterile liquid; 100mL; Composition: Dextran/Sodium Diatrizoate; 1.086 +/- 0.001g/cu. cm at 22 deg.C; 6.9 +/- 0.3 pH; Storage: 4 deg.C, Protect from light
Catalog No. NC9025455 Supplier Cederlane Supplier No. CL5115
Lympholyte;-Mammal; Sterile liquid; 100mL; Composition: Dextran/Sodium Diatrizoate; 1.086 +/- 0.001g/cu. cm at 22 deg.C; 6.9 +/- 0.3 pH; Storage: 4 deg.C, Protect from light