Learn More
Cederlane LYMPHOLYTE-POLY STERILE LIQUI
SDP
Catalog No. NC9177930
Encompass
Lympholyte�-poly Cell Separation Media (Isolation of PMNs from Human peripheral blood), sterile liquid, Size: 100ml
Catalog No. NC9177930 Supplier Cederlane Supplier No. CL5070
Lympholyte�-poly Cell Separation Media (Isolation of PMNs from Human peripheral blood), sterile liquid, Size: 100ml