Learn More
Essentra Porous Technologies FG Clean Wipes Presaturated Wipes Canister, 6" x 8.5" Wipes, 96% IPA/4% DI Water, 100 Wipes/Canister, 12 Canisters/CS
SDP
Catalog No. NC1474511
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements. The FG Clean Wipes 6-LS964-685 wipe removes dust, dirt, oxides, and most contaminants from electronic equipment and most surfaces. Wipes are made with strong, durable hydro-entangled fabric that will not tear easily. They are handy, convenient, economical and disposable. Single-use wipes are perfect for cleaning work stations, counter tops, door knobs, conference tables, telephones, surfaces and more. These wipes are perfect for cleaning plastic pipe prior to electrofusion. They are widely used by utility companies.
This item is not returnable. View return policy
Catalog No. NC1474511 Supplier Essentra Porous Technologies Supplier No. 6LS964685
This item has a minimum order quantity of 3 per supplier requirements. The FG Clean Wipes 6-LS964-685 wipe removes dust, dirt, oxides, and most contaminants from electronic equipment and most surfaces. Wipes are made with strong, durable hydro-entangled fabric that will not tear easily. They are handy, convenient, economical and disposable. Single-use wipes are perfect for cleaning work stations, counter tops, door knobs, conference tables, telephones, surfaces and more. These wipes are perfect for cleaning plastic pipe prior to electrofusion. They are widely used by utility companies.