Learn More
Selleck Chemical LLC LYN-1604 2mg 2088939-99-3
SDP
Supplier:  Selleck Chemical LLC S85972MG
Encompass_Preferred
LYN-1604 is a potential ULK1 agonist with IC50 of 1.66 ?M against MDA-MB-231 cells and it binds to wild-type ULK1 with a binding affinity in the nanomole range (Kd=291.4 nM). The ULK1 (Y89A) mutant protein caused a sharp decrease in binding affinity with lower response and Kd than wild-type ULK1, ULK1 (K50A) and ULK1 (L53A) mutants. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8223