Learn More
TARGETMOL CHEMICALS INC LYN-1604 dihydrochloride 10MG
SDP
Catalog No. 502877035
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.LYN-1604 is a novel ULK1 activator.It inducing cell death involved in ATF3, RAD21, and caspase3, accompanied by autophagy and apoptosis. Purity 99.8%
Catalog No. 50-287-7035 Supplier TARGETMOL CHEMICALS INC Supplier No. T8808L10MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.LYN-1604 is a novel ULK1 activator.It inducing cell death involved in ATF3, RAD21, and caspase3, accompanied by autophagy and apoptosis. Purity 99.8%