Learn More
Medchemexpress LLC Lyn-1604 dihydrochloride | 2310109-38-5 | 99.6% | 657.54 | C33H45Cl4N3O2 | 100 MG
SDP
Catalog No. 5000248227
Encompass_Preferred
LYN-1604 dihydrochloride is a research-grade small molecule activator of UNC-51-like kinase 1 (ULK1), used to study autophagy and cell death pathways in preclinical models such as triple negative breast cancer. The compound is supplied as the dihydrochloride salt and is provided with analytical documentation for research use.
  • Potent ULK1 activator (EC50 = 18.94 nM).
  • High purity suitable for research applications.
  • Available as solid and as a DMSO solution for convenient dosing.
  • Analytical documentation available, including COA, HNMR, and LCMS.
  • Supplied as the dihydrochloride salt for consistent formulation.
Catalog No. 50-002-48227 Supplier Medchemexpress LLC Supplier No. HY101923B100MG
LYN-1604 dihydrochloride is a research-grade small molecule activator of UNC-51-like kinase 1 (ULK1), used to study autophagy and cell death pathways in preclinical models such as triple negative breast cancer. The compound is supplied as the dihydrochloride salt and is provided with analytical documentation for research use.
  • Potent ULK1 activator (EC50 = 18.94 nM).
  • High purity suitable for research applications.
  • Available as solid and as a DMSO solution for convenient dosing.
  • Analytical documentation available, including COA, HNMR, and LCMS.
  • Supplied as the dihydrochloride salt for consistent formulation.