Learn More
Medchemexpress LLC Lyn-1604 dihydrochloride | 2310109-38-5 | 99.6% | 657.54 g·mol-1 | C33H45Cl4N3O2 | 10 MG
SDP
Catalog No. 5000248340
Encompass_Preferred
LYN-1604 dihydrochloride is a small-molecule activator of UNC-51-like kinase 1 (ULK1) used to study autophagy and triple-negative breast cancer. Supplied as the dihydrochloride salt, it is provided at high purity for in vitro biochemical and cellular assays.
  • Activates ULK1 with nanomolar EC50 (18.94 nM).
  • High purity solid suitable for biochemical and cellular research.
  • Available as small-mass solids and pre-made DMSO solution for convenience.
  • Stable when stored sealed at 4°C; in solvent store at -80°C for long term.
  • Dihydrochloride salt form improves solubility for assay preparation.
Catalog No. 50-002-48340 Supplier Medchemexpress LLC Supplier No. HY101923B10MG
LYN-1604 dihydrochloride is a small-molecule activator of UNC-51-like kinase 1 (ULK1) used to study autophagy and triple-negative breast cancer. Supplied as the dihydrochloride salt, it is provided at high purity for in vitro biochemical and cellular assays.
  • Activates ULK1 with nanomolar EC50 (18.94 nM).
  • High purity solid suitable for biochemical and cellular research.
  • Available as small-mass solids and pre-made DMSO solution for convenience.
  • Stable when stored sealed at 4°C; in solvent store at -80°C for long term.
  • Dihydrochloride salt form improves solubility for assay preparation.