Learn More
Medchemexpress LLC Lyn-1604 dihydrochloride | 2310109-38-5 | 99.6% | 657.54 g/mol | C33H45Cl4N3O2 | 25 MG
SDP
Catalog No. 5000245725
Encompass_Preferred
LYN-1604 dihydrochloride is a potent small-molecule activator of UNC-51-like kinase 1 (ULK1) used in preclinical research, including studies of triple-negative breast cancer. The compound exhibits strong activity (reported EC50 18.94 nM) and is supplied as the dihydrochloride salt to improve handling and solubility. For research use only.
  • Potent ULK1 activation (EC50 18.94 nM).
  • High purity (≈99.6%).
  • Supplied as a dihydrochloride salt for improved solubility.
  • Solid form; stable when stored sealed at 4°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month) per guidance.
  • Available in multiple small research pack sizes suitable for preclinical studies.
Catalog No. 50-002-45725 Supplier Medchemexpress LLC Supplier No. HY101923B25MG
LYN-1604 dihydrochloride is a potent small-molecule activator of UNC-51-like kinase 1 (ULK1) used in preclinical research, including studies of triple-negative breast cancer. The compound exhibits strong activity (reported EC50 18.94 nM) and is supplied as the dihydrochloride salt to improve handling and solubility. For research use only.
  • Potent ULK1 activation (EC50 18.94 nM).
  • High purity (≈99.6%).
  • Supplied as a dihydrochloride salt for improved solubility.
  • Solid form; stable when stored sealed at 4°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month) per guidance.
  • Available in multiple small research pack sizes suitable for preclinical studies.