Learn More
Medchemexpress LLC Lyn-1604 dihydrochloride | 2310109-38-5 | 99.6% | 657.54 g/mol | C33H45Cl4N3O2 | 50 MG
SDP
Catalog No. 5000248869
Encompass_Preferred
LYN-1604 dihydrochloride is a small-molecule activator of UNC-51-like kinase 1 (ULK1) intended for biochemical and cellular research into autophagy and triple-negative breast cancer. The compound activates ULK1 (EC50 = 18.94 nM), shows cellular potency (IC50 = 1.66 μM in MDA-MB-231 cells), and has a measured binding affinity (KD = 291.4 nM). Supplied as the dihydrochloride salt at high purity for assay and dose-response studies.
  • Potent ULK1 activator (EC50 = 18.94 nM).
  • Demonstrated cellular activity (IC50 = 1.66 μM in MDA-MB-231 cells).
  • Measured binding affinity to ULK1 (KD = 291.4 nM).
  • High purity suitable for biochemical and cellular assays (99.55%).
  • Available in multiple sizes for dose-range and scale-up studies.
Catalog No. 50-002-48869 Supplier Medchemexpress LLC Supplier No. HY101923B50MG
LYN-1604 dihydrochloride is a small-molecule activator of UNC-51-like kinase 1 (ULK1) intended for biochemical and cellular research into autophagy and triple-negative breast cancer. The compound activates ULK1 (EC50 = 18.94 nM), shows cellular potency (IC50 = 1.66 μM in MDA-MB-231 cells), and has a measured binding affinity (KD = 291.4 nM). Supplied as the dihydrochloride salt at high purity for assay and dose-response studies.
  • Potent ULK1 activator (EC50 = 18.94 nM).
  • Demonstrated cellular activity (IC50 = 1.66 μM in MDA-MB-231 cells).
  • Measured binding affinity to ULK1 (KD = 291.4 nM).
  • High purity suitable for biochemical and cellular assays (99.55%).
  • Available in multiple sizes for dose-range and scale-up studies.