Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LYN-1604 hydrochloride | 2216753-86-3 | MFCD31630830 | >98.0%
SDP

Catalog No. 5000246519
Encompass_Preferred

LYN-1604 hydrochloride | 2216753-86-3 | MFCD31630830 | >98.0%

Catalog No. 50-002-46519 Supplier Medchemexpress LLC Supplier No. HY101923A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LYN-1604 hydrochloride | 2216753-86-3 | MFCD31630830 | >98.0%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.