Learn More
Medchemexpress LLC LYN-1604 hydrochloride | 2216753-86-3 | MFCD31630830 | >=98.0% | 621.08 | C33H44Cl3N3O2 | 1 MG
SDP
Catalog No. 5000248759
Encompass_Preferred
A small-molecule hydrochloride salt that activates UNC-51-like kinase 1 (ULK1) with a reported EC50 of 18.94 nM. It is used in vitro to study autophagy, apoptosis, and mechanisms relevant to triple-negative breast cancer. Supplied as a solid and as a 10 mM solution in DMSO; consult the certificate of analysis for lot-specific purity and storage instructions.
  • Potent ULK1 activation (EC50 = 18.94 nM).
  • Supports studies of autophagy and apoptosis in cancer models.
  • Available as solid and as a 10 mM solution in DMSO for assay flexibility.
  • Hydrochloride salt form improves solubility for biological testing.
  • Certificate of analysis available for lot-specific purity and storage instructions.
Catalog No. 50-002-48759 Supplier Medchemexpress LLC Supplier No. HY101923A1MG
A small-molecule hydrochloride salt that activates UNC-51-like kinase 1 (ULK1) with a reported EC50 of 18.94 nM. It is used in vitro to study autophagy, apoptosis, and mechanisms relevant to triple-negative breast cancer. Supplied as a solid and as a 10 mM solution in DMSO; consult the certificate of analysis for lot-specific purity and storage instructions.
  • Potent ULK1 activation (EC50 = 18.94 nM).
  • Supports studies of autophagy and apoptosis in cancer models.
  • Available as solid and as a 10 mM solution in DMSO for assay flexibility.
  • Hydrochloride salt form improves solubility for biological testing.
  • Certificate of analysis available for lot-specific purity and storage instructions.