Learn More
Sino Biological Recombinant Mouse LMTK3 Protein (GST Tag)
Catalog No. 502574477
Encompass_Preferred
Recombinant mouse LMTK3 (63-484) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4477 Supplier Sino Biological Supplier No. L0835G100
Recombinant mouse LMTK3 (63-484) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.