Learn More
Aviva Systems Biology Corp Lyn Antibody - N-terminal region (ARP54697_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54697_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Lyn. It was validated on Western Blot. . Alternate names: AA407514, Hck-2, p53Lyn, p56LynApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-956