Learn More
Aviva Systems Biology Corp LYN Antibody - N-terminal region (ARP54696_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54696_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LYN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LYN. Alternate names: FLJ26625, JTK8, p53Lyn, p56LynApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-62986