Learn More
TARGETMOL CHEMICALS INC LYN-IN-1 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1191610MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lyn-IN-1 (Bafetinib analog) is a selective and potent dual inhibitor of Bcr-Abl and Lynpurity: 99%
Catalog No. 50-257-7436