Learn More
Medchemexpress LLC Lyn peptide inhibitor | 222018-18-0 | 99.35% | 2370.88 | 25 MG
SDP
Catalog No. 5000426992
Encompass_Preferred
Lyn peptide inhibitor is a potent and cell-permeable inhibitor of the Lyn-coupled IL-5 receptor signaling pathway, designed to keep other signals intact. It works by blocking Lyn activation and inhibiting the binding of Lyn tyrosine kinase to the βc subunit of IL-3/GM-CSF/IL-5 receptors. This inhibitor is suitable for studying asthma, allergic, and other eosinophilic disorders.
  • Potent and cell-permeable inhibitor
  • Blocks Lyn activation
  • Inhibits binding of Lyn tyrosine kinase to the βc subunit of IL-3/GM-CSF/IL-5 receptors
  • Can be used for the study of asthma, allergic, and other eosinophilic disorders
  • Targets IL-5 and IL-3
  • Inhibits Src and Interleukin related pathways
  • Involved in Protein Tyrosine Kinase/RTK and Immunology/Inflammation pathways
  • Sequence: {Stearoyl}-Tyr-Gly-Tyr-Arg-Leu-Arg-Arg-Lys-Trp-Glu-Glu-Lys-Ile-Pro-Asn-Pro-NH2
Catalog No. 50-004-26992 Supplier Medchemexpress LLC Supplier No. 25MGHYP1111
Lyn peptide inhibitor is a potent and cell-permeable inhibitor of the Lyn-coupled IL-5 receptor signaling pathway, designed to keep other signals intact. It works by blocking Lyn activation and inhibiting the binding of Lyn tyrosine kinase to the βc subunit of IL-3/GM-CSF/IL-5 receptors. This inhibitor is suitable for studying asthma, allergic, and other eosinophilic disorders.
  • Potent and cell-permeable inhibitor
  • Blocks Lyn activation
  • Inhibits binding of Lyn tyrosine kinase to the βc subunit of IL-3/GM-CSF/IL-5 receptors
  • Can be used for the study of asthma, allergic, and other eosinophilic disorders
  • Targets IL-5 and IL-3
  • Inhibits Src and Interleukin related pathways
  • Involved in Protein Tyrosine Kinase/RTK and Immunology/Inflammation pathways
  • Sequence: {Stearoyl}-Tyr-Gly-Tyr-Arg-Leu-Arg-Arg-Lys-Trp-Glu-Glu-Lys-Ile-Pro-Asn-Pro-NH2