Learn More
Medchemexpress LLC Lyn peptide inhibitor | 222018-18-0 | 99.4% | 2370.88 | 10 MG
SDP
Catalog No. 5000424178
Encompass_Preferred
Lyn peptide inhibitor is a potent and cell-permeable inhibitor of the Lyn-coupled IL-5 receptor signaling pathway. It blocks Lyn activation and inhibits the binding of Lyn tyrosine kinase to the βc subunit of IL-3/GM-CSF/IL-5 receptors. This inhibitor is useful for the study of asthma, allergic, and other eosinophilic disorders.
  • Potent and cell-permeable inhibitor
  • Blocks Lyn activation
  • Inhibits binding of Lyn tyrosine kinase to βc subunit of IL-3/GM-CSF/IL-5 receptors
  • White to off-white solid appearance
  • Soluble in DMSO: 5 mg/mL (2.11 mM; requires ultrasonic)
Catalog No. 50-004-24178 Supplier Medchemexpress LLC Supplier No. HYP111110MG
Add to Cart
Add to Cart
Lyn peptide inhibitor is a potent and cell-permeable inhibitor of the Lyn-coupled IL-5 receptor signaling pathway. It blocks Lyn activation and inhibits the binding of Lyn tyrosine kinase to the βc subunit of IL-3/GM-CSF/IL-5 receptors. This inhibitor is useful for the study of asthma, allergic, and other eosinophilic disorders.
  • Potent and cell-permeable inhibitor
  • Blocks Lyn activation
  • Inhibits binding of Lyn tyrosine kinase to βc subunit of IL-3/GM-CSF/IL-5 receptors
  • White to off-white solid appearance
  • Soluble in DMSO: 5 mg/mL (2.11 mM; requires ultrasonic)