Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lyn peptide inhibitor | 222018-18-0 | 99.4% | 2370.88 | 5 MG
SDP

Catalog No. 5000422715
Encompass_Preferred

Lyn peptide inhibitor is a potent and cell-permeable inhibitor of the Lyn-coupled IL-5 receptor signaling pathway, while keeping other signals intact. It blocks Lyn activation and inhibits the binding of Lyn tyrosine kinase to the βc subunit of IL-3/GM-CSF/IL-5 receptors. It can be used for the study of asthma, allergic, and other eosinophilic disorders.

  • Potent and cell-permeable inhibitor
  • Blocks Lyn activation
  • Inhibits binding of Lyn tyrosine kinase to the βc subunit of IL-3/GM-CSF/IL-5 receptors
  • Used for the study of asthma
  • Used for the study of allergic disorders
  • Used for the study of other eosinophilic disorders
  • Appearance: White to off-white solid
  • Powder storage: -80°C for 2 years, -20°C for 1 year (sealed, away from moisture)
  • In solvent storage: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)
  • Soluble in DMSO (5 mg/mL)

Catalog No. 50-004-22715 Supplier Medchemexpress LLC Supplier No. HYP11115MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lyn peptide inhibitor is a potent and cell-permeable inhibitor of the Lyn-coupled IL-5 receptor signaling pathway, while keeping other signals intact. It blocks Lyn activation and inhibits the binding of Lyn tyrosine kinase to the βc subunit of IL-3/GM-CSF/IL-5 receptors. It can be used for the study of asthma, allergic, and other eosinophilic disorders.

  • Potent and cell-permeable inhibitor
  • Blocks Lyn activation
  • Inhibits binding of Lyn tyrosine kinase to the βc subunit of IL-3/GM-CSF/IL-5 receptors
  • Used for the study of asthma
  • Used for the study of allergic disorders
  • Used for the study of other eosinophilic disorders
  • Appearance: White to off-white solid
  • Powder storage: -80°C for 2 years, -20°C for 1 year (sealed, away from moisture)
  • In solvent storage: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)
  • Soluble in DMSO (5 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.