Learn More
Enzo Life Sciences Lyn & Syk kinase substrate (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP2170001
Encompass_Preferred
This peptide, EDPIYEFLPAKKK, was identified through phage display as a high affinity substrate for both Lyn kinase (Km=24 µM) and Syk kinase (Km=17 µM). The three lysines on the C-terminus allow this peptide to be used in phosphocellulose kinase assays. Purity: ≥95%. Long Term Storage: -20°C.
Catalog No. 50-201-1292