Learn More
Carnabio Usa Inc LYNA/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 081715
Encompass
LYNa, Full-length, Wild type, Amino Acid 1-512, NP_002341.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0488