Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lynestrenol | 52-76-6 | 99.87% | 284.44 | 25 MG
SDP

Catalog No. 5000347621
Encompass_Preferred

Lynestrenol is a pregnane X receptor (PXR) agonist. It is a synthetic progesteronelike agent that possesses immunostimulatory properties. It enhances lymphocyte response to phytohemagglutinin, mixed lymphocyte culture, active T-rosette test, autologous red cell rosettes, and increases monocyte phagocytosis.

  • Purity: 99.87%
  • Molecular weight: 284.44
  • Formula: C20H28O
  • CAS No.: 52-76-6
  • Appearance: Solid
  • Color: White to off-white
  • For research use only

Catalog No. 50-003-47621 Supplier Medchemexpress LLC Supplier No. 25MGHY109568
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lynestrenol is a pregnane X receptor (PXR) agonist. It is a synthetic progesteronelike agent that possesses immunostimulatory properties. It enhances lymphocyte response to phytohemagglutinin, mixed lymphocyte culture, active T-rosette test, autologous red cell rosettes, and increases monocyte phagocytosis.

  • Purity: 99.87%
  • Molecular weight: 284.44
  • Formula: C20H28O
  • CAS No.: 52-76-6
  • Appearance: Solid
  • Color: White to off-white
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.