Learn More
EMD MILLIPORE 3P BIOPHARM LYNX ST CONNECTOR 1/2 IN. NON-
Catalog No. NC9689066
Encompass
1/2 inch Lynx ST Connector, Enables integration of steamable hard piped process equipment with disposable sterile fluid paths
Catalog No. NC9689066 Supplier EMD MILLIPORE 3P BIOPHARM Supplier No. STC21THN01
1/2 inch Lynx ST Connector, Enables integration of steamable hard piped process equipment with disposable sterile fluid paths