Learn More
Aviva Systems Biology Corp Lynx1 Antibody - middle region (ARP58708_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58708_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Lynx1. It was validated on Western Blot. . Alternate names: AI838844ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-286-492