Learn More
Meridian Life Science Lyo-Compatible MMLV-RT, 80
SDP
Catalog No. 5000856229
Encompass_Preferred
Lyo-Compatible MMLV-RT is a high-concentration, Moloney Murine Leukemia Virus (MMLV) Reverse Transcriptase (RT) with reduced RNase H activity, which exhibits high sensitivity and efficiency for cDNA synthesis and is suitable for incorporation into wet or lyophilized multiplex RT-PCR assays, allowing high sensitivity detection of low copy number RNA targets.
Catalog No. 50-008-56229 Supplier Meridian Life Science Supplier No. MDX042008
Lyo-Compatible MMLV-RT is a high-concentration, Moloney Murine Leukemia Virus (MMLV) Reverse Transcriptase (RT) with reduced RNase H activity, which exhibits high sensitivity and efficiency for cDNA synthesis and is suitable for incorporation into wet or lyophilized multiplex RT-PCR assays, allowing high sensitivity detection of low copy number RNA targets.